missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10590552
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10590552 250 mg 250mg
10374322 1 g 1g
10727285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10590552 Supplier Thermo Scientific Maybridge Supplier No. CC18203CB
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 215434-25-6
Molecular Formula C8H6ClNO2S3
Molecular Weight (g/mol) 279.771
InChI Key YIKNRCVJBOLZFL-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonyl chloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-sulphonyl chloride, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulphonylchloride, 5-2-methyl-thiazol-4-yl-thiophene-2-sulfonyl chloride, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenesulfonyl chloride, chloro 5-2-methyl 1,3-thiazol-4-yl 2-thienyl sulfone
PubChem CID 2736979
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride
SMILES CC1=NC(=CS1)C2=CC=C(S2)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 279.79
Percent Purity 95%
Chemical Name or Material 5-(2-Methyl-1, 3-thiazol-4-yl)thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.