missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Phenyleth-1-ynyl)-2-furoic acid, Tech., Thermo Scientific™

Product Code. 10753063
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10753063 250 mg 250mg
10049261 1 g 1g
10652895 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10753063 Supplier Thermo Scientific Maybridge Supplier No. KM08625CB
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 130423-83-5
Molecular Formula C13H8O3
Molecular Weight (g/mol) 212.20
MDL Number MFCD00541949
InChI Key HFYACYZCOUYIFR-UHFFFAOYSA-N
Synonym 5-phenylethynyl furan-2-carboxylic acid, 5-2-phenyleth-1-ynyl-2-furoic acid, 5-2-phenylethynyl furan-2-carboxylic acid, 5-phenylethynyl-2-furoic acid, 2-furancarboxylic acid,5-2-phenylethynyl, maybridge1_008982, acmc-20a65w, 5-phenylethynyl-2-furoic acid, tech, 5-phenylethynyl-furan 2 carboxylic acid
PubChem CID 2737138
IUPAC Name 5-(2-phenylethynyl)furan-2-carboxylic acid
SMILES OC(=O)C1=CC=C(O1)C#CC1=CC=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.2
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)-2-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.