missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde, 90%, Thermo Scientific™

Product Code. 10702503
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10702503 10 g Amber Glass Bottle 10g
10394192 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702503 Supplier Thermo Scientific Maybridge Supplier No. KM08144EA
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 17257-10-2
Molecular Formula C13H8OS
Molecular Weight (g/mol) 212.266
MDL Number MFCD00126374
InChI Key YFMUACLZRVJOBK-UHFFFAOYSA-N
Synonym 5-2-phenyleth-1-ynyl thiophene-2-carbaldehyde, 5-phenylethynyl thiophene-2-carbaldehyde, 5-2-phenylethynyl thiophene-2-carbaldehyde, 5-phenylethynyl thiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde,5-2-phenylethynyl, 5-phenylethynyl-thiophene-2-carbaldehyde, 5-phenylethynyl thiophene-2-carboxylate
PubChem CID 2737140
IUPAC Name 5-(2-phenylethynyl)thiophene-2-carbaldehyde
SMILES C1=CC=C(C=C1)C#CC2=CC=C(S2)C=O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 212.27
Percent Purity 90%
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.