missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Pyridinyl)-2-thiophenecarbothioamide, 97%, Thermo Scientific™

Product Code. 10723803
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10723803 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10723803 Supplier Thermo Scientific Maybridge Supplier No. CC00077DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 306934-91-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H8N2S2
MDL Number MFCD02677740
Quantity 1 g
Synonym 5-2-pyridinyl-2-thiophenecarbothioamide, 5-pyridin-2-yl thiophene-2-carbothioamide, 2-thiophenecarbothioamide,5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-thiocarboxamide, amino 5-2-pyridyl 2-thienyl methane-1-thione
InChI Key VOYHCOULKKDLRP-UHFFFAOYSA-N
SMILES C1=CC=NC(=C1)C2=CC=C(S2)C(=S)N
IUPAC Name 5-pyridin-2-ylthiophene-2-carbothioamide
Molecular Weight (g/mol) 220.308
PubChem CID 2776127
Formula Weight 220.32
Percent Purity 97%
Chemical Name or Material 5-(2-Pyridinyl)-2-thiophenecarbothioamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.