missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Pyridyl)thiophene-2-sulfonyl chloride, Technical Grade, Thermo Scientific™

Product Code. 10742283
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10742283 1 g 1g
10776705 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10742283 Supplier Thermo Scientific Maybridge Supplier No. CC02203DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 151858-64-9
Molecular Formula C9H6ClNO2S2
Molecular Weight (g/mol) 259.72
MDL Number MFCD00052284
InChI Key OGOMWPWAEIDEOU-UHFFFAOYSA-N
Synonym 5-2-pyridyl thiophene-2-sulfonyl chloride, 5-pyridin-2-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, 5-pyridin-2-yl thiophene-2-sulphonyl chloride, tech, acmc-1c5sp, 5-pyrid-2-ylthiophene-2-sulfonyl chloride, chloro 5-2-pyridyl 2-thienyl sulfone, 2-5-chlorosulphonyl thien-2-yl pyridine, 5-2-pyridyl thiophene-2-sulphonyl chloride
PubChem CID 2737231
IUPAC Name 5-pyridin-2-ylthiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 259.74
Percent Purity 95%
Chemical Name or Material 5-(2-Pyridyl)thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.