missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Thienyl)-3-isoxazolyl]methanol, 97%, Thermo Scientific™

Product Code. 10590732
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10590732 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10590732 Supplier Thermo Scientific Maybridge Supplier No. CC23109CB
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 194491-44-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H7NO2S
MDL Number MFCD03086162
Quantity 250 mg
Synonym 5-2-thienyl-3-isoxazolyl methanol, 5-thiophen-2-yl isoxazol-3-yl methanol, 5-thiophen-2-yl-1,2-oxazol-3-yl methanol, 3-isoxazolemethanol,5-2-thienyl, 5-thiophen-2-ylisoxazol-3-yl methanol, 3mq, 5-2-thienyl isoxazol-3-yl methanol, 5-2-thienyl isoxazol-3-yl methan-1-ol, 5-thiophen-2-yl isoxazol-3-yl methanol .
InChI Key HUAGDHXVPCSWLD-UHFFFAOYSA-N
SMILES C1=CSC(=C1)C2=CC(=NO2)CO
IUPAC Name (5-thiophen-2-yl-1,2-oxazol-3-yl)methanol
Molecular Weight (g/mol) 181.209
PubChem CID 2776547
Formula Weight 181.21
Percent Purity 97%
Chemical Name or Material [5-(2-Thienyl)-3-isoxazolyl]methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.