missing translation for 'onlineSavingsMsg'
Learn More

5-[2-(Trifluoromethoxy)phenyl]-2-furaldehyde, Thermo Scientific™

Product Code. 10712133
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712133 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10712133 Supplier Thermo Scientific Maybridge Supplier No. SEW05184DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 306936-00-5
Packaging Amber Glass Bottle
Molecular Formula C12H7F3O3
MDL Number MFCD00278715
Quantity 1 g
Synonym 5-2-trifluoromethoxy phenyl-2-furaldehyde, 5-2-trifluoromethoxy phenyl furan-2-carbaldehyde, 2-furancarboxaldehyde,5-2-trifluoromethoxy phenyl, cbmicro_046447, regid_for_cid_721838, 5-2-trifluoromethoxy-phenyl-furan-2-carbaldehyde, 5-2-trifluoromethoxy phenyl furan-2-carboxaldehyde, 2-furancarboxaldehyde, 5-2-trifluoromethoxy phenyl
InChI Key UTGWIIGCXFJJHZ-UHFFFAOYSA-N
SMILES FC(F)(F)OC1=CC=CC=C1C1=CC=C(O1)C=O
IUPAC Name 5-[2-(trifluoromethoxy)phenyl]furan-2-carbaldehyde
Molecular Weight (g/mol) 256.18
PubChem CID 721838
Formula Weight 256.18
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-[2-(Trifluoromethoxy)phenyl]-2-furaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.