missing translation for 'onlineSavingsMsg'
Learn More

5-[2-(Trifluoromethyl)phenyl]-1,2-oxazole-4-carboxylic Acid, TRC

Product Code. 30456455
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456455 10 mg -
30449041 100 mg 100mg
30456428 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456455 Supplier Toronto Research Chemicals Supplier No. TRCT79212510MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003561-80-5
Molecular Formula C11H6NO3F3
Molecular Weight (g/mol) 257.17
IUPAC Name 5-[2-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxylic acid
SMILES OC(=O)c1cnoc1c2ccccc2C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H6F3NO3/c12-11(13,14)8-4-2-1-3-6(8)9-7(10(16)17)5-15-18-9/h1-5H,(H,16,17)
Formula Weight 257.03
Chemical Name or Material 5-[2-(Trifluoromethyl)phenyl]-1,2-oxazole-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.