missing translation for 'onlineSavingsMsg'
Learn More

5-(3,4-Dichlorophenyl)furan-2-carbonyl chloride, Thermo Scientific™

Product Code. 11345389
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345389 1 g 1g
11335389 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345389 Supplier Thermo Scientific Alfa Aesar Supplier No. H50394.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61941-91-1
Molecular Formula C11H5Cl3O2
Molecular Weight (g/mol) 275.51
MDL Number MFCD02257993
InChI Key XQRLANCABKJNFO-UHFFFAOYSA-N
Synonym 5-3,4-dichlorophenyl furan-2-carbonyl chloride, 2-furancarbonyl chloride, 5-3,4-dichlorophenyl, 5-3,4-dichloro-phenyl-furan-2-carbonyl chloride
PubChem CID 3873501
IUPAC Name 5-(3,4-dichlorophenyl)furan-2-carbonyl chloride
SMILES ClC(=O)C1=CC=C(O1)C1=CC(Cl)=C(Cl)C=C1

Specifications

Quantity 1 g
UN Number UN3261
Formula Weight 275.52
Chemical Name or Material 5-(3,4-Dichlorophenyl)furan-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.