missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Isocyanatophenyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10618544
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10618544 1 g Amber Glass Bottle 1g
10154893 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10618544 Supplier Thermo Scientific Maybridge Supplier No. CC60506DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 941716-86-5
Molecular Formula C11H9N3O
Molecular Weight (g/mol) 199.213
MDL Number MFCD09817502
InChI Key ZFRFBJVFEQIMLM-UHFFFAOYSA-N
Synonym 5-3-isocyanatophenyl-1-methyl-1h-pyrazole, 5-3-isocyanatophenyl-1-methylpyrazole, 3-1-methylpyrazol-5-yl benzenisocyanate, 3-1-methyl-1h-pyrazol-5-yl phenylisocyanate
PubChem CID 24229624
IUPAC Name 5-(3-isocyanatophenyl)-1-methylpyrazole
SMILES CN1C(=CC=N1)C2=CC(=CC=C2)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 5-(3-Isocyanatophenyl)-1-methyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.