missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Pyridyl)-1H-tetrazole, 98%

Product Code. 11494487
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11494487

Brand: Thermo Scientific Alfa Aesar B20492.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3250-74-6 | C6H5N5 | 147.141 g/mol

5-(3-Pyridyl)-1H-tetrazole is used as a pharmaceutical intermediate. The Pyridine,3-(2H-tetrazol-5-yl)- acts as reactant with 1-(4-bromo-phenyl)-3-piperidino-propan-1-one, hydrochloride, and obtain the 1-(4-bromo-phenyl)-3-(5-pyridin-3-yl-tetrazol-2-yl)-propan-1-one. This reaction needs the solvent of dimethylformamide. The yield is 31%. In addition, this reaction should be taken for 8 hours at the temperature of 150°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(3-Pyridyl)-1H-tetrazole is used as a pharmaceutical intermediate. The Pyridine,3-(2H-tetrazol-5-yl)- acts as reactant with 1-(4-bromo-phenyl)-3-piperidino-propan-1-one, hydrochloride, and obtain the 1-(4-bromo-phenyl)-3-(5-pyridin-3-yl-tetrazol-2-yl)-propan-1-one. This reaction needs the solvent of dimethylformamide. The yield is 31%. In addition, this reaction should be taken for 8 hours at the temperature of 150°C.

Solubility
Soluble in water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 3250-74-6
Molecular Formula C6H5N5
Molecular Weight (g/mol) 147.141
MDL Number MFCD00040281
InChI Key SECHDFHDDVELCV-UHFFFAOYSA-N
Synonym 3-2h-tetrazol-5-yl pyridine, 5-3-pyridyl-1h-tetrazole, 3-1h-tetrazol-5-yl pyridine, 5-3-pyridyl tetrazole, 3-2h-tetrazol-5-yl-pyridine, pyridine, 3-2h-tetrazol-5-yl, unii-zc84pps834, 3-2h-1,2,3,4-tetrazol-5-yl pyridine, 3-1h-1,2,3,4-tetrazol-5-yl pyridine, 5-3-pyridyl-2h-1,2,3,4-tetraazole
PubChem CID 151091
IUPAC Name 3-(2H-tetrazol-5-yl)pyridine
SMILES C1=CC(=CN=C1)C2=NNN=N2

Especificaciones

Melting Point ∼240°C (decomposition)
Odor Odorless
Quantity 1 g
Solubility Information Soluble in water (slightly).
Formula Weight 147.14
Percent Purity 98%
Chemical Name or Material 5-(3-Pyridyl)-1H-tetrazole

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado