missing translation for 'onlineSavingsMsg'
Learn More

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran, 97%, Thermo Scientific™

Product Code. 10502413
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10502413 1 g 1g
11300567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10502413 Supplier Thermo Scientific Maybridge Supplier No. CC07139DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 519054-55-8
Molecular Formula C14H17BO3
Molecular Weight (g/mol) 244.10
MDL Number MFCD04115374
InChI Key ZQCCCOMKYKVFFN-UHFFFAOYSA-N
Synonym 2-benzofuran-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-benzofuran, benzofuran-5-boronic acid pinacol ester, 2-1-benzofuran-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, benzo b furan-5-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzo b furan, benzofuran, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzofuran, pubchem20151, 5-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl-1-benzofuran
PubChem CID 2795190
IUPAC Name 2-(1-benzofuran-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C2OC=CC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 244.1
Percent Purity 97%
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.