missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol, 97%, Thermo Scientific™

Product Code. 10261363
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10261363 1 g 1g
10792937 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10261363 Supplier Thermo Scientific Maybridge Supplier No. TG00105DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 17452-31-2
Molecular Formula C10H9ClN2S
Molecular Weight (g/mol) 224.71
MDL Number MFCD01313844
InChI Key RGYNOPLNIDBIGT-UHFFFAOYSA-N
Synonym 5-4-chlorophenyl-1-methyl-1h-imidazole-2-thiol, 5-4-chlorophenyl-1-methylimidazole-2-thiol, maybridge1_000141, 4-4-chlorophenyl-3-methyl-1h-imidazole-2-thione, 5-4-chlorophenyl-1-methyl-3h-imidazole-2-thione, 2h-imidazole-2-thione,5-4-chlorophenyl-1,3-dihydro-1-methyl
PubChem CID 2747520
IUPAC Name 4-(4-chlorophenyl)-3-methyl-1H-imidazole-2-thione
SMILES CN1C(=S)NC=C1C1=CC=C(Cl)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 224.71
Chemical Name or Material 5-(4-Chlorophenyl)-1-methyl-1H-imidazole-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.