missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)-2-methyl-3-furoic acid, ≥97%, Thermo Scientific™

Product Code. 10538523
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10538523 10 g 10g
10583053 1 g 1g
10086434 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10538523 Supplier Thermo Scientific Maybridge Supplier No. SP00065EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 111787-89-4
Molecular Formula C12H9ClO3
Molecular Weight (g/mol) 236.65
MDL Number MFCD00275560
InChI Key HUQOLZUHGMRQFS-UHFFFAOYSA-N
PubChem CID 2742181
IUPAC Name 5-(4-chlorophenyl)-2-methylfuran-3-carboxylic acid
SMILES CC1=C(C=C(O1)C1=CC=C(Cl)C=C1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Synonym 5-4-chlorophenyl-2-methylfuran-3-carboxylic acid, 5-4-chlorophenyl-2-methyl-3-furoic acid, 2-methyl-5-4-chlorophenyl-furan-3-carboxylic acid, 3-furancarboxylic acid,5-4-chlorophenyl-2-methyl, maybridge1_008585, acmc-20met1, 5-4-chlorophenyl-2-methyl-3-furancarboxylic acid, 3-furancarboxylic acid, 5-4-chlorophenyl-2-methyl
Formula Weight 236.65
Percent Purity 97%
Chemical Name or Material 5-(4-Chlorophenyl)-2-methyl-3-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.