missing translation for 'onlineSavingsMsg'
Learn More

5-(4-chlorophenyl)-2-(trifluoromethyl)-3-furoic acid, 97%, Thermo Scientific™

Product Code. 10539782
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10539782 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10539782 Supplier Thermo Scientific Maybridge Supplier No. SP00061DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175276-60-5
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H6ClF3O3
MDL Number MFCD00275555
Quantity 1 g
Synonym 5-4-chlorophenyl-2-trifluoromethyl furan-3-carboxylic acid, 5-4-chlorophenyl-2-trifluoromethyl-3-furoic acid, 3-furancarboxylic acid,5-4-chlorophenyl-2-trifluoromethyl, maybridge1_008497, pubchem7362, 5-4-chlorophenyl-2-trifluoromethyl-3-furancarboxylic acid
InChI Key BQUIEVOFWHVZAW-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C2=CC(=C(O2)C(F)(F)F)C(=O)O)Cl
IUPAC Name 5-(4-chlorophenyl)-2-(trifluoromethyl)furan-3-carboxylic acid
Molecular Weight (g/mol) 290.622
PubChem CID 2736644
Formula Weight 290.62
Percent Purity 97%
Chemical Name or Material 5-(4-Chlorophenyl)-2-(trifluoromethyl)-3-furoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.