missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Dimethylaminobenzylidene)rhodanine 98%

Product Code. 11403377
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11403377 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403377 Supplier Thermo Scientific Alfa Aesar Supplier No. A17152.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 536-17-4 | C12H12N2OS2 | 264.36 g/mol

Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applied for the detection of Ag, Au, Cu, Hg, Pd, Pt, alkaloids, antipyrin, indican, sulfonamide, urobilinogen. Reagent for the determination of silver, palladium, mercury and gold. Also used in the photometric determination of auto radiograms.

Solubility
Insoluble in water, 993.7(mg/L) at 25°C

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 536-17-4
Melting Point 275°C to 280°C
Odor Weak
Molecular Formula C12H12N2OS2
MDL Number MFCD00064857
Quantity 10 g
Beilstein 189065
Merck Index 14,3231
Synonym unii-0 ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0 ser53q7rt, usaf pd-20
Solubility Information Insoluble aq. soln.,993.7(mg/L) at 25°C
InChI Key JJRVRELEASDUMY-YFHOEESVSA-N
SMILES CN(C)C1=CC=C(\C=C2/SC(=S)NC2=O)C=C1
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol) 264.36
PubChem CID 2723826
Formula Weight 264.37
Percent Purity 98%
Chemical Name or Material 5-(4-Dimethylaminobenzylidene)rhodanine 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.