missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Methylphenyl)-5-phenylhydantoin, 99+%, Thermo Scientific™

Product Code. 10665612 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10665612 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10665612 Supplier Thermo Scientific Acros Supplier No. 171430010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51169-17-6
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.29
MDL Number MFCD00005263
InChI Key WPAPSGQWYNPWCZ-UHFFFAOYSA-N
Synonym 5-p-methylphenyl-5-phenylhydantoin, 5-phenyl-5-p-tolyl hydantoin, 5-phenyl-5-4-tolyl hydantoin, 5-4-methylphenyl-5-phenylimidazolidine-2,4-dione, 5-4-methylphenyl-5-phenyl-2,4-imidazolidinedione, 5-p-tolyl-5-phenylhydantoin, 5-phenyl-5-p-tolyl imidazolidine-2,4-dione, 2,4-imidazolidinedione, 5-4-methylphenyl-5-phenyl, 5-4-methylphenyl-5-phenylhydantoin, 5-4-methylphenyl-5-phenyl hydantoin
PubChem CID 92334
IUPAC Name 5-(4-methylphenyl)-5-phenylimidazolidine-2,4-dione
SMILES CC1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 225.0°C to 228.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 1 g
Formula Weight 266.29
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 5-(4-Methylphenyl)-5-phenylhydantoin, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.