missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Nitrophenyl)-1H-tetrazole, 97%

Product Code. 11337476
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11337476 1 g 1g
11347476 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11337476

Brand: Thermo Scientific Alfa Aesar L09531.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16687-60-8 | C7H5N5O2 | 191.15 g/mol

It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.

Solubility
Insoluble in water. Soluble in DMSO.

Notes
 It may be stored in the dark in a desiccator. Avoid contact with strong heat and metal salts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16687-60-8
Molecular Formula C7H5N5O2
Molecular Weight (g/mol) 191.15
MDL Number MFCD00068729
InChI Key MIUOBAHGBPSRKY-UHFFFAOYSA-N
Synonym 5-4-nitrophenyl-1h-tetrazole, 5-4-nitrophenyl-2h-tetrazole, 5-4-nitro-phenyl-2h-tetrazole, 5-4-nitrophenyl tetrazole, 5-4-nitrophenyl-1h-1,2,3,4-tetrazole, 5-4-nitro-phenyl-1h-tetrazole, 5-4-nitrophenyl-2h-1,2,3,4-tetrazole, maybridge1_004022, acmc-1bt31
PubChem CID 285163
IUPAC Name 5-(4-nitrophenyl)-2H-tetrazole
SMILES [O-][N+](=O)C1=CC=C(C=C1)C1=NNN=N1

Specifications

Melting Point ∽223°C (decomposition)
Quantity 1 g
Beilstein 192840
Solubility Information Insoluble in water. Soluble in DMSO.
Formula Weight 191.15
Percent Purity 97%
Chemical Name or Material 5-(4-Nitrophenyl)-1H-tetrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.