missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Nitrophenyl)-1H-tetrazole, 97%

Product Code. 11347476
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347476 5 g 5g
11337476 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347476 Supplier Thermo Scientific Alfa Aesar Supplier No. L09531.06
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16687-60-8 | C7H5N5O2 | 191.15 g/mol

It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.

Solubility
Insoluble in water. Soluble in DMSO.

Notes
 It may be stored in the dark in a desiccator. Avoid contact with strong heat and metal salts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16687-60-8
Molecular Formula C7H5N5O2
Molecular Weight (g/mol) 191.15
MDL Number MFCD00068729
InChI Key MIUOBAHGBPSRKY-UHFFFAOYSA-N
Synonym 5-4-nitrophenyl-1h-tetrazole, 5-4-nitrophenyl-2h-tetrazole, 5-4-nitro-phenyl-2h-tetrazole, 5-4-nitrophenyl tetrazole, 5-4-nitrophenyl-1h-1,2,3,4-tetrazole, 5-4-nitro-phenyl-1h-tetrazole, 5-4-nitrophenyl-2h-1,2,3,4-tetrazole, maybridge1_004022, acmc-1bt31
PubChem CID 285163
IUPAC Name 5-(4-nitrophenyl)-2H-tetrazole
SMILES [O-][N+](=O)C1=CC=C(C=C1)C1=NNN=N1

Specifications

Melting Point ∽223°C (decomposition)
Quantity 5 g
Beilstein 192840
Solubility Information Insoluble in water. Soluble in DMSO.
Formula Weight 191.15
Percent Purity 97%
Chemical Name or Material 5-(4-Nitrophenyl)-1H-tetrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.