missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Pyridyl)-1,3,4-oxadiazole-2-thiol, 97%, Thermo Scientific™

Product Code. 15485677
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15485677

Brand: Thermo Scientific Alfa Aesar H32907.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15264-63-8
Molecular Formula C7H5N3OS
Molecular Weight (g/mol) 179.20
MDL Number MFCD00215200
InChI Key TXCXZVFDWQYTIC-UHFFFAOYSA-N
Synonym 5-4-pyridyl-1,3,4-oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4-oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4 oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4-oxadiazole-2 3h-thione, 5-4-pyridinyl-1,3,4-oxadiazole-2-thiol, 1,3,4-oxadiazole-2-thiol, 5-4-pyridinyl, 1,3,4-oxadiazole-2 3h-thione,5-4-pyridinyl, 5-pyridin-4-yl-3h-1,3,4-oxadiazole-2-thione, d04qyn
PubChem CID 774383
SMILES S=C1NN=C(O1)C1=CC=NC=C1

Specifications

Melting Point 262°C to 264°C
Quantity 5 g
IUPAC Name 5-(pyridin-4-yl)-2,3-dihydro-1,3,4-oxadiazole-2-thione
Formula Weight 179.2
Percent Purity 97%
Chemical Name or Material 5-(4-Pyridyl)-1,3,4-oxadiazole-2-thiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.