missing translation for 'onlineSavingsMsg'
Learn More

5-[5-(2-Thienyl)-2-thienyl]thiophene-2-carbaldehyde, Thermo Scientific™

Product Code. 10474643
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10474643 1 g 1g
10577363 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10474643 Supplier Thermo Scientific Maybridge Supplier No. SEW04053DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7342-41-8
Molecular Formula C13H8OS3
Molecular Weight (g/mol) 276.39
MDL Number MFCD00115181
InChI Key PMPDDPJYARBNGV-UHFFFAOYSA-N
Synonym 2,2':5',2-terthiophene-5-carboxaldehyde, 5-5-2-thienyl-2-thienyl thiophene-2-carbaldehyde, 2,2':5',2-terthiophene-5-carbaldehyde, 5-formyl-2,2':5',2-terthiophene, chembl91933, 5-5-thiophen-2-ylthiophen-2-yl thiophene-2-carbaldehyde, 5-5-thiophen-2-yl thiophen-2-yl thiophene-2-carbaldehyde, 2-formyl-alpha-terthienyl
PubChem CID 454742
SMILES O=CC1=CC=C(S1)C1=CC=C(S1)C1=CC=CS1

Specifications

Quantity 1 g
IUPAC Name 5'-(thiophen-2-yl)-[2,2'-bithiophene]-5-carbaldehyde
Formula Weight 276.4
Chemical Name or Material 5-[5-(2-Thienyl)-2-thienyl]thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.