missing translation for 'onlineSavingsMsg'
Learn More

5-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10438022
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10438022 250 mg Amber Glass Bottle 250mg
10683613 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10438022 Supplier Thermo Scientific Maybridge Supplier No. SPB08412CB
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 306937-21-3
Molecular Formula C6H2Cl2N2O2S3
Molecular Weight (g/mol) 301.174
MDL Number MFCD02180888
InChI Key DCLCBWFFYZNVES-UHFFFAOYSA-N
Synonym 5-5-chloro-1,2,4-thiadiazol-3-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-5-chloro-1,2,4-thiadiazol-3-yl, 2-5-chloro-1,2,4-thiadiazol-3-yl thiophene-5-sulfonyl chloride, 5-5-chloro-1,2,4-thiadiazol-3-yl-2-thiophenesulfonyl chloride, 5-5-chloro-1,2,4-thiadiazol-3-yl-thiophene-2-sulfonyl chloride
PubChem CID 2735820
IUPAC Name 5-(5-chloro-1,2,4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1)S(=O)(=O)Cl)C2=NSC(=N2)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 301.2
Percent Purity 95%
Chemical Name or Material 5-(5-Chloro-1, 2, 4-thiadiazol-3-yl)thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.