missing translation for 'onlineSavingsMsg'
Learn More

5,5'-Dibromo-2,2'-bithiophene, 98%

Product Code. 15422808
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422808 25 g 25g
15412808 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422808 Supplier Thermo Scientific Alfa Aesar Supplier No. H56337.14
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4805-22-5 | C8H4Br2S2 | 324.048 g/mol

It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4805-22-5
Molecular Formula C8H4Br2S2
Molecular Weight (g/mol) 324.048
MDL Number MFCD00219110
InChI Key SXNCMLQAQIGJDO-UHFFFAOYSA-N
Synonym 5,5'-dibromo-2,2'-bithiophene, 2-bromo-5-5-bromothiophen-2-yl thiophene, 5,5-dibromo-2,2-bithiophene, 2,2'-bithiophene, 5,5'-dibromo, 5,5'-bis 2-bromothiophene, 5,5'-dibromo-2,2'bithiophene, 5,5'-dibromo-2,2-bithiophene, maybridge4_002009, acmc-209kc4, ksc491i4r
PubChem CID 638908
IUPAC Name 2-bromo-5-(5-bromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1)Br)C2=CC=C(S2)Br

Specifications

Melting Point 144°C to 146°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 324.06
Percent Purity 99%
Chemical Name or Material 5,5'-Dibromo-2,2'-bithiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.