missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethyl-1,3-cyclohexanedione, 99%

Product Code. 10666682 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10666682

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126-81-8 | C8H12O2 | 140.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-81-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001588
InChI Key BADXJIPKFRBFOT-UHFFFAOYSA-N
Synonym dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
PubChem CID 31358
IUPAC Name 5,5-dimethylcyclohexane-1,3-dione
SMILES CC1(C)CC(=O)CC(=O)C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 147.0°C to 150.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2C6H6(=O)2
Quantity 500 g
Beilstein 07, 559
Fieser 01,266
Merck Index 15, 3264
Solubility Information Solubility in water: 5 % (25°C). Other solubilities: soluble in methanol,ethanol,benzene,chloroform,and acetic acid
Formula Weight 140.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5, 5-Dimethyl-1, 3-cyclohexanedione, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.