missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethyl-1,3-dioxane-2-butanal, 96%

Product Code. 11484907
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484907 2.5 g 2.5g
11494907 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484907 Supplier Thermo Scientific Alfa Aesar Supplier No. B21858.88
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 127600-13-9 | C10H18O3 | 186.251 g/mol

It is employed as a intermediate for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceuticals.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127600-13-9
Molecular Formula C10H18O3
Molecular Weight (g/mol) 186.251
MDL Number MFCD02683076
InChI Key KSKRSPYEXSAZRE-UHFFFAOYSA-N
Synonym 4-5,5-dimethyl-1,3-dioxan-2-yl butanal, 5,5-dimethyl-1,3-dioxane-2-butanal, glutaraldehyde mono-neopentyl glycol acetal, 5,5-dimethyl-2-3-formylpropyl-1,3-dioxane, 5,5-dimethyl-2-3'-formylpropyl-1,3-dioxane
PubChem CID 2734769
IUPAC Name 4-(5,5-dimethyl-1,3-dioxan-2-yl)butanal
SMILES CC1(COC(OC1)CCCC=O)C

Specifications

Refractive Index 1.449
Quantity 2.5 g
Sensitivity Air Sensitive
Formula Weight 186.25
Percent Purity 96%
Chemical Name or Material 5,5-Dimethyl-1,3-dioxane-2-butanal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.