missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dimethylhydantoin, TRC

Product Code. 30387716
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387716 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387716 Supplier Toronto Research Chemicals Supplier No. TRCD4733005G
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,5-Dimethylhydantoin
CAS 77-71-4
Molecular Formula C5H8N2O2
Quantity 5 g
Synonym 5,5-Dimethyl-2,4-imidazolidinedione, 4,4-Dimethyl-2,5-dioxoimidazolidine, 5,5-Dimethylhydantoin, DM Hydantoin, DMH, Dantoin 736, Dantoin DMH, Dimethylhydantoin, Fennosurf 300, NSC 8652
InChI Formula InChI=1S/C5H8N2O2/c1-5(2)3(8)6-4(9)7-5/h1-2H3,(H2,6,7,8,9)
SMILES CC1(C)NC(=O)NC1=O
Molecular Weight (g/mol) 128.13
Formula Weight 128.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.