missing translation for 'onlineSavingsMsg'
Learn More

5,5-Dioxide Thioridazine Disulfoxide, TRC

Product Code. 30137493
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137493 100 mg 100mg
30135176 10 mg 10mg
30011105 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137493 Supplier Toronto Research Chemicals Supplier No. TRCD100575100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100574-22-9
Molecular Formula C21 H26 N2 O4 S2
Molecular Weight (g/mol) 434.57
Synonym 10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylsulfonyl)-10H-phenothiazine 5,5-Dioxide, Sulforidazine 5-Sulfone, Thioridazine Hydrochloride Imp. A (EP), Thioridazine Imp. A (EP), Thioridazine Hydrochloride Impurity A, Thioridazine Impurity A
IUPAC Name 10-[2-(1-methylpiperidin-2-yl)ethyl]-2-methylsulfonylphenothiazine 5,5-dioxide
SMILES CN1CCCCC1CCN2c3ccccc3S(=O)(=O)c4ccc(cc24)S(=O)(=O)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H26N2O4S2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)29(26,27)21-11-10-17(15-19(21)23)28(2,24)25/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3
Formula Weight 434.1334
Chemical Name or Material Sulforidazine 5-Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.