missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin, 99%

Product Code. 11468866
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468866 100 g 100g
11458866 25 g 25g
11334679 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468866 Supplier Thermo Scientific Alfa Aesar Supplier No. A17013.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57-41-0 | C15H12N2O2 | 252.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 57-41-0
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
MDL Number MFCD00005264
InChI Key CXOFVDLJLONNDW-UHFFFAOYSA-N
Synonym phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan
PubChem CID 1775
ChEBI CHEBI:8107
IUPAC Name 5,5-diphenylimidazolidine-2,4-dione
SMILES C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3

Specifica

Melting Point 293°C to 298°C
Quantity 100 g
Beilstein 384532
Merck Index 14,7322
Formula Weight 252.27
Percent Purity 99%
Chemical Name or Material 5,5-Diphenylhydantoin
compliance-icons compliance-icons
Identificativo prodotto
  • 5,5-Diphenylhydantoin
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
Indicazioni di pericolo
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
Consigli di prudenza
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Informazioni supplementari
  • MIXTURE LIST-Contain: Phenytoin

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.