missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin, 99+%, Thermo Scientific Chemicals

Product Code. 10791061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10791061 100 g Plastic bottle 100g
10042180 5 g Glass bottle 5g
10676572 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10791061 Supplier Thermo Scientific Acros Supplier No. 171731000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-41-0
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.27
MDL Number MFCD00005264
InChI Key CXOFVDLJLONNDW-UHFFFAOYSA-N
Synonym phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan
PubChem CID 1775
ChEBI CHEBI:8107
IUPAC Name 5,5-diphenylimidazolidine-2,4-dione
SMILES C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 293.5°C to 295°C
Color White
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Quantity 100 g
Beilstein 24, 410
Merck Index 15, 7433
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alkali hydroxides,1g/60mL alcohol,1g/30mL acetone
Formula Weight 252.27
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 5, 5-Diphenylhydantoin, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5,5-Diphenylhydantoin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.