missing translation for 'onlineSavingsMsg'
Learn More

5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10672833
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10672833 250 mg 250mg
10232842 1 g 1g
10366804 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10672833 Supplier Thermo Scientific Maybridge Supplier No. KM09559CB
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 229956-98-3
Molecular Formula C8H3ClF3NO3S2
Molecular Weight (g/mol) 317.681
MDL Number MFCD00831001
InChI Key TWGONVLMWWEOFL-UHFFFAOYSA-N
Synonym 5-5-trifluoromethyl isoxazol-3-yl thiophene-2-sulfonyl chloride, 5-5-trifluoromethyl-1,2-oxazol-3-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-5-trifluoromethyl-3-isoxazolyl, 5-5-trifluoromethyl isoxazol-3-yl thiophene-2-sulphonyl chloride, 5-5-trifluoromethyl-isoxazol-3-yl-thiophene-2-sulfonyl chloride
PubChem CID 2777526
IUPAC Name 5-[5-(trifluoromethyl)-1,2-oxazol-3-yl]thiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1)S(=O)(=O)Cl)C2=NOC(=C2)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 317.69
Percent Purity 97%
Chemical Name or Material 5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.