missing translation for 'onlineSavingsMsg'
Learn More

5(6)-Dehydro-4(5)-dihydro D-(-)-Norgestrel (>90%), TRC

Product Code. 30364730
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364730 25 mg 25mg
30350085 10 mg 10mg
30354289 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364730 Supplier Toronto Research Chemicals Supplier No. TRCD22993525MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100021-05-4
Molecular Formula C21 H28 O2
Molecular Weight (g/mol) 312.45
Synonym Levonorgestrel Imp. P (EP), 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-5-en-20-yn-3-one (δ5-Levonorgestrel)
IUPAC Name (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-17-hydroxy-1,2,4,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
SMILES CC[C@]12CC[C@H]3[C@@H](CC=C4CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,5,16-19,23H,3,6-13H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1
Formula Weight 312.2089
Chemical Name or Material 5(6)-Dehydro-4(5)-dihydro D-(-)-Norgestrel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.