missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydro Tofacitinib, TRC

Product Code. 30134367
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134367 50 mg 50mg
30010509 25 mg 25mg
30007968 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134367 Supplier Toronto Research Chemicals Supplier No. TRCD45024050MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1640972-35-5
Molecular Formula C16H22N6O
Molecular Weight (g/mol) 314.39
Synonym 5,6-Dihydro CP-690550, 3-((3R,4R)-3-((6,7-dihydro-5H-pyrrolo[2,3-d]pyrimidin-4-yl)(methyl)amino)-4-methylpiperidin-1-yl)-3-oxopropanenitrile
SMILES O=C(CC#N)N(C1)CC[C@@H](C)[C@H]1N(C)C2=C3C(NCC3)=NC=N2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H22N6O/c1-11-5-8-22(14(23)3-6-17)9-13(11)21(2)16-12-4-7-18-15(12)19-10-20-16/h10-11,13H,3-5,7-9H2,1-2H3,(H,18,19,20)/t11-,13+/m1/s1
Formula Weight 314.1855
Percent Purity >95%
Chemical Name or Material 5,6-Dihydro CP-690550
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.