missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dihydrouridine, MedChemExpress

Product Code. 16437467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437467 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16437467 Supplier Medchem Express Supplier No. HY113047
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5,6-Dihydrouridine is a modified base found in conserved positions in the D-loop of tRNA in Bacteria, Eukaryota, and some Archaea.

5,6-Dihydrouridine is a modified base found in conserved positions in the D-loop of tRNA in Bacteria, Eukaryota, and some Archaea.

Specifications

Chemical Name or Material 5,6-Dihydrouridine
CAS 5627-05-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C9H14N2O6
Quantity 5 mg
Solubility Information H2O : 100 mg/mL (406.14 mM; Need ultrasonic) ∣DMSO : 35.71 mg/mL (145.03 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2CCC(NC2=O)=O)O1)O)O
Molecular Weight (g/mol) 246.22
Formula Weight 246.22
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.