missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethoxy-2-[(4-piperidyl)methyl]indane Hydrochloride(Donepezil Impurity), TRC

Product Code. 30370331
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370331 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370331 Supplier Toronto Research Chemicals Supplier No. TRCD4607215MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,6-Dimethoxy-2-[(4-piperidyl)methyl]indane Hydrochloride(Donepezil Impurity)
CAS 1034439-43-4
Molecular Formula C17H26ClNO2
Quantity 5 mg
Synonym 4-[(2,3-Dihydro-5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine Hydrochloride, Donepezil Desbenzyl Deoxy Impurity F
InChI Formula InChI=1S/C17H25NO2.ClH/c1-19-16-10-14-8-13(7-12-3-5-18-6-4-12)9-15(14)11-17(16)20-2;/h10-13,18H,3-9H2,1-2H3;1H
SMILES Cl.COc1cc2CC(CC3CCNCC3)Cc2cc1OC
Molecular Weight (g/mol) 311.85
Formula Weight 311.1652
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.