Learn More
5,6-Dimethoxyindole-2-carboxylic acid, 98%
Description
5,6-Dimethoxyindole-2-carboxylic acid is used as a reagent and heterocyclic building blocks in synthetic chemistry. It is used to prepare a melanin-like polymer, poly(5,6-dimethoxyindole-2-carboxylic acid), which exhibits electrochromic properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5,6-Dimethoxyindole-2-carboxylic acid is used as a reagent and heterocyclic building blocks in synthetic chemistry. It is used to prepare a melanin-like polymer, poly(5,6-dimethoxyindole-2-carboxylic acid), which exhibits electrochromic properties.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Spécification
Spécification
| Melting Point | ∽210°C (decomposition) |
| Quantity | 5 g |
| Beilstein | 202249 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 221.22 |
| Percent Purity | 98% |
| Chemical Name or Material | 5,6-Dimethoxyindole-2-carboxylic acid |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.