missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, 97+%

Product Code. 11322896
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322896 5 g 5g
11312896 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322896 Supplier Thermo Scientific Alfa Aesar Supplier No. L10607.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1264198-47-1 | C20H12N4Na2O6S2 | 514.44 g/mol

Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264198-47-1
Molecular Formula C20H12N4Na2O6S2
Molecular Weight (g/mol) 514.44
MDL Number MFCD00717621
InChI Key DDIHLFCSEHAOJZ-UHFFFAOYSA-L
Synonym ferrozin, ferrozine, ferrozine disodium salt, unii-29j2ciw9pk, 29j2ciw9pk, 3-2-pyridyl-5,6-di 4-phenylsulfonic acid-1,2,4-triazine, disodium salt, 4,4'-3-2-pyridyl-triazine-5,6-diyl dibenzenesulfonic acid disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid, disodium salt, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-benzenesulfonic acid,disodium salt, benzenesulfonic acid, 4,4'-3-2-pyridinyl-1,2,4-triazine-5,6-diyl bis-, disodium salt
PubChem CID 34127
IUPAC Name disodium 4-[3-(pyridin-2-yl)-5-(4-sulfonatophenyl)-1,2,4-triazin-6-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=C(C=C1)S([O-])(=O)=O)C1=CC=CC=N1

Specifications

Quantity 5 g
Beilstein 5222087
Solubility Information Soluble in water.
Formula Weight 514.46 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate
compliance-icons
Product Identifier
  • 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.