missing translation for 'onlineSavingsMsg'
Learn More

5,6-Methylenedioxy-2-aminoindane, TRC

Product Code. 30479824
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479824 500 mg 500mg
30472280 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30479824 Supplier Toronto Research Chemicals Supplier No. TRCM303990500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132741-81-2
Molecular Formula C10 H11 N O2
Molecular Weight (g/mol) 177.2
Synonym MDAI, 5,6-Methylenedioxy-2-aminoindane
IUPAC Name 6,7-dihydro-5H-cyclopenta[f][1,3]benzodioxol-6-amine
SMILES NC1Cc2cc3OCOc3cc2C1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H11NO2/c11-8-1-6-3-9-10(13-5-12-9)4-7(6)2-8/h3-4,8H,1-2,5,11H2
Formula Weight 177.079
Chemical Name or Material 5,6-Methylenedioxy-2-aminoindane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.