missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dimethylisatin, 95%, ACROS Organics™

Product Code. 10613562 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10613562 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10613562 Supplier Thermo Scientific Acros Supplier No. 338740010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 39603-24-2
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C10H9NO2
MDL Number MFCD00047219
Quantity 1 g
Synonym 5,7-dimethylisatin, 5,7-dimethylindoline-2,3-dione, 5,7-dimethyl-indole-2,3-dione, 5,7-dimethyl-2,3-dihydro-1h-indole-2,3-dione, 1h-indole-2,3-dione, 5,7-dimethyl, indole-2,3-dione, 5,7-dimethyl, 1h-indole-2,3-dione, 5,7-dimethyl-9ci, acmc-20a4zq, isatin-based compound, 48, 1h-indole-2,3-dione,5,7-dimethyl
InChI Key HFZSCCJTJGWTDZ-UHFFFAOYSA-N
SMILES CC1=CC(=C2C(=C1)C(=O)C(=O)N2)C
IUPAC Name 5,7-dimethyl-1H-indole-2,3-dione
Molecular Weight (g/mol) 175.187
PubChem CID 38296
Formula Weight 175.19
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material 5, 7-Dimethylisatin, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.