missing translation for 'onlineSavingsMsg'
Learn More

5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine, 95%, Thermo Scientific Chemicals

Product Code. 15436068
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15436068 1 g 1g
15495186 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436068 Supplier Thermo Scientific Alfa Aesar Supplier No. H59681.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 959755-46-5 | C5H2Br2N4 | 277.907 g/mol

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959755-46-5
Molecular Formula C5H2Br2N4
Molecular Weight (g/mol) 277.907
MDL Number MFCD11040378
InChI Key IDWOTLNDEITXRA-UHFFFAOYSA-N
Synonym 5,8-dibromo-1,2,4 triazolo 1,5-a pyrazine, 5,8-dibromo 1,2,4 triazolo 1,5-a pyrazine, 5,8-dibromo-1,2,4-triazolo 1,5-a pyrazine, 5,8-dibromo 1,2,4 triazolo 1,5-a-pyrazine
PubChem CID 49758022
IUPAC Name 5,8-dibromo-[1,2,4]triazolo[1,5-a]pyrazine
SMILES C1=C(N2C(=NC=N2)C(=N1)Br)Br

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 277.91
Percent Purity 95%
Chemical Name or Material 5,8-Dibromo-1,2,4-triazolo[1,5-a]pyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.