missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-amino-4-methylthiazole, 97+%

Product Code. 11387226
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387226 5 g 5g
11397226 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387226 Supplier Thermo Scientific Alfa Aesar Supplier No. L09108.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 30748-47-1 | C6H8N2OS | 156.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30748-47-1
Molecular Formula C6H8N2OS
Molecular Weight (g/mol) 156.203
MDL Number MFCD00051952
InChI Key PKUKCASRNJIQNU-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-5-acetylthiazole, 1-2-amino-4-methylthiazol-5-yl ethanone, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethanone, 5-acetyl-2-amino-4-methylthiazole, 1-2-amino-4-methyl-1,3-thiazol-5-yl ethan-1-one, 1-2-amino-4-methyl-thiazol-5-yl-ethanone, 5-acetyl-2-amino-4-methyl-1,3-thiazole, ethanone, 1-2-amino-4-methyl-5-thiazolyl, 1-2-amino-4-methyl-1,3-thiazol-5-yl-1-ethanone, 5-acetyl-4-methyl-2-thiazolamine
PubChem CID 720882
IUPAC Name 1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)N)C(=O)C

Specifications

Melting Point ∽268°C (decomposition)
Quantity 5 g
Beilstein 127111
Formula Weight 156.21
Percent Purity ≥97%
Chemical Name or Material 5-Acetyl-2-amino-4-methylthiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.