missing translation for 'onlineSavingsMsg'
Learn More

5-(Acetylamino)-N,N’-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, TRC

Product Code. 30319051
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319051 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319051 Supplier Toronto Research Chemicals Supplier No. TRCA16818050MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 5-(Acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
CAS 31127-80-7
Molecular Formula C16 H20 I3 N3 O7
Quantity 50 mg
Synonym 5-(Acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide, 5-(Acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide, 5-Acetylamino-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide, 5-Acetamido-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-isophthalamide, Iohexol impurity A (EP), Iohexol Related Compound A (USP)
InChI Formula InChI=1S/C16H20I3N3O7/c1-6(25)22-14-12(18)9(15(28)20-2-7(26)4-23)11(17)10(13(14)19)16(29)21-3-8(27)5-24/h7-8,23-24,26-27H,2-5H2,1H3,(H,20,28)(H,21,29)(H,22,25)
SMILES CC(=O)Nc1c(I)c(C(=O)NCC(O)CO)c(I)c(C(=O)NCC(O)CO)c1I
IUPAC Name 5-acetamido-1-N,3-N-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular Weight (g/mol) 747.06
Formula Weight 746.8435
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.