missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylindane, 97%

Product Code. 11419406
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
This item is not returnable. View return policy

Product Code. 11419406

Brand: Thermo Scientific Alfa Aesar A19102.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4228-10-8 | C11H12O | 160.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4228-10-8
MDL Number MFCD00021246

Specifications

Boiling Point 85°C to 87°C (0.05 mmHg)
Molecular Formula C11H14O3
Quantity 50 g
Synonym isobutyl 4-hydroxybenzoate, isobutylparaben, isobutyl p-hydroxybenzoate, isobutyl paraben, isobutyl-4-hydroxybenzoate, isobutyl parahydroxybenzoate, 4-hydroxybenzoic acid isobutyl ester, benzoic acid, 4-hydroxy-, 2-methylpropyl ester, p-oxybenzoesaureisobutylester, unii-0qqj25x58g
InChI Key XPJVKCRENWUEJH-UHFFFAOYSA-N
SMILES CC(C)COC(=O)C1=CC=C(C=C1)O
IUPAC Name 2-methylpropyl 4-hydroxybenzoate
Molecular Weight (g/mol) 194.23
PubChem CID 20240
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material 5-Acetylindane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.