missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1,3,4-thiadiazole-2-thiol, TRC

Product Code. 30240984
Change view
Click to view available options
Quantity:
1 g
100 g
Unit Size:
100g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240984 1 g 1g
30240919 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240984 Supplier Toronto Research Chemicals Supplier No. TRCA6333001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2349-67-9
Molecular Formula C2H3N3S2
Molecular Weight (g/mol) 133.19
Synonym 1,3,4-Thiadiazole-2-thiol, 5-amino- (6CI,8CI), δ2-1,3,4-Thiadiazoline-2-thiol, 5-imino- (7CI), 5-Amino-1,3,4-thiadiazole-2(3H)-thione, 2-Amino-1,3,4-thiadiazol-5-thiol, 2-Amino-1,3,4-thiadiazole-5-mercaptan, 2-Amino-1,3,4-thiadiazole-5-thiol, 2-Amino-5-mercapto-1,3,4-thiadiazole, 2-Amino-5-mercaptothiadiazole, 2-Amino-5-sulfanyl-1,3,4-thiadiazole, 2-Amino-5-sulfhydryl-1,3,4-thiadiazole, 2-Amino-δ2-1,3,4-thiadiazoline-5-thione, 2-Mercapto-5-amino-1,3,4-thiadiazole, 2-Mercapto-5-aminothiadiazole, 2-Thiol-5-amino-1,3,4-thiadiazole, 5-Amino-1,3,4-thiadiazole-2-thiol, 5-Amino-1,3,4-thiadiazole-2-thione, 5-Amino-1,3,4-thiadiazole-5-thiol, 5-Amino-1,3,4-thiadiazoline-2-thione, 5-Amino-1,3,4-thidiazole-2-thiol, 5-Amino-2-mercapto-1,3,4-thiadiazole, 5-Amino-2-sulfhydryl-1,3,4-thiadiazole, 5-Amino-3H-1,3,4-thiadiazole-2-thione, 5-Thio-2-amino-[1,3,4]thiadiazole, 5-Thiol-2-amino-1,3,4-thiadiazole, ATT, NSC 209061, NSC 21402, WR 180
IUPAC Name 5-amino-1,3,4-thiadiazole-2-thiol
SMILES Nc1nnc(S)s1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H3N3S2/c3-1-4-5-2(6)7-1/h(H2,3,4)(H,5,6)
Formula Weight 132.9768
Chemical Name or Material 5-Amino-1,3,4-thiadiazole-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.