missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific™

Product Code. 10600903
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10600903 10 g Amber Glass Bottle 10g
10232392 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10600903 Supplier Thermo Scientific Maybridge Supplier No. BTB07485EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51516-67-7
Molecular Formula C10H7ClN4
Molecular Weight (g/mol) 218.64
MDL Number MFCD00052031
InChI Key WIWSALMJHPGFDY-UHFFFAOYSA-N
Synonym 5-amino-1-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile, 5-amino-1-4-chlorophenyl, maybridge1_001852, acmc-1b1ld, 5-amino-1-4-chloro-phenyl-1h-pyrazole-4-carbonit, 1-4-chlorophenyl-5-amino-1h-pyrazole-4-carbonitrile
PubChem CID 639097
SMILES NC1=C(C=NN1C1=CC=C(Cl)C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Quantity 10 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
IUPAC Name 5-amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Formula Weight 218.65
Percent Purity 97%
Chemical Name or Material 5-Amino-1-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.