missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-methyl-1H-indazole, 97%, Thermo Scientific™

Product Code. 10760540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10760540 5 g Glass bottle 5g
10309747 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10760540 Supplier Thermo Scientific Acros Supplier No. 429520050
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 50593-24-3
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
MDL Number MFCD03305455
InChI Key PYOFNPHTKBSXOM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazol-5-amine, 5-amino-1-methyl-1h-indazole, 1-methyl-1h-indazol-5-ylamine, 1h-indazol-5-amine, 1-methyl, 5-amino-1-methylindazole, 1-methyl-1h-indazole-5-amine, pubchem20587, 1-methyl-5-aminoindazol, 1-methyl-5-indazolamine, 1-methyl-5-aminoindazole
PubChem CID 2768032
IUPAC Name 1-methylindazol-5-amine
SMILES CN1C2=C(C=C(C=C2)N)C=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 154.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 147.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1-methyl-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-1-methyl-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.