missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-methyl-1H-pyrazole-4-carboxamide, Thermo Scientific™

Product Code. 10763053
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10763053 250 mg Amber Glass Bottle 250mg
10600283 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10763053 Supplier Thermo Scientific Maybridge Supplier No. KM00775CB
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 18213-75-7
Molecular Formula C5H8N4O
Molecular Weight (g/mol) 140.146
MDL Number MFCD00111808
InChI Key JGSQVTVXGXOSCH-UHFFFAOYSA-N
PubChem CID 265696
IUPAC Name 5-amino-1-methylpyrazole-4-carboxamide
SMILES CN1C(=C(C=N1)C(=O)N)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-amino-1-methyl-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide, 5-amino-1-methyl, pubchem22392, 5-amino-1-methyl-4-pyrazolecarboxamide, 5-azanyl-1-methyl-pyrazole-4-carboxamide, 1-methyl-5-amino-1h-pyrazole-4-carboxamide, 1h-pyrazole-4-carboxamide,5-amino-1-methyl, 5-imino-1-methyl-2h-pyrazole-4-carboxamide, 3-imino-2-methyl-2,3-dihydro-1h-pyrazole-4-carboxamide
Formula Weight 140.15
Percent Purity 95%
Chemical Name or Material 5-Amino-1-methyl-1H-pyrazole-4-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.