missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-pentanol, TRC

Product Code. 30133444
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133444 100 g 100g
30010702 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133444 Supplier Toronto Research Chemicals Supplier No. TRCA618975100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2508-29-4
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.16
Synonym 1-Amino-5-hydroxypentane, 1-Amino-5-pentanol, 5-Aminopentanol, 5-Hydroxy-1-pentanamine, 5-Hydroxypentylamine, Pentanolamine;
IUPAC Name 5-aminopentan-1-ol
SMILES NCCCCCO

Specifications

Quantity 100 g
InChI Formula InChI=1S/C5H13NO/c6-4-2-1-3-5-7/h7H,1-6H2
Formula Weight 103.0997
Chemical Name or Material 5-Amino-1-pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.