missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1-(tert-butyl)-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile, 97%, Thermo Scientific™

Product Code. 10120992
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10120992 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10120992 Supplier Thermo Scientific Maybridge Supplier No. SS00047DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 180903-14-4
CAS Min % 97.0
Quantity 1 g
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C14H15ClN4
MDL Number MFCD01935935
Synonym 5-amino-1-tert-butyl-3-4-chlorophenyl-1h-pyrazole-4-carbonitrile, 5-amino-1-tert-butyl-3-4-chlorophenyl pyrazole-4-carbonitrile, 1h-pyrazole-4-carbonitrile,5-amino-3-4-chlorophenyl-1-1,1-dimethylethyl, aminobutylchlorophenylpyrazolecarbonitrile, 1-tert-butyl-3-4-chlorophenyl-5-amino-1h-pyrazole-4-carbonitrile
InChI Key HKCFCHLGYDCCSP-UHFFFAOYSA-N
SMILES CC(C)(C)N1C(=C(C(=N1)C2=CC=C(C=C2)Cl)C#N)N
IUPAC Name 5-amino-1-tert-butyl-3-(4-chlorophenyl)pyrazole-4-carbonitrile
Molecular Weight (g/mol) 274.752
PubChem CID 2747447
Formula Weight 274.75
Percent Purity 97%
Chemical Name or Material 5-Amino-1-(tert-butyl)-3-(4-chlorophenyl)-1H-pyrazole-4-carbonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.