missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2,2-difluoro-1,3-benzodioxole, 97+%

Product Code. 11435767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435767 5 g 5g
11425767 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435767 Supplier Thermo Scientific Alfa Aesar Supplier No. B24336.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1544-85-0 | C7H5F2NO2 | 173.119 g/mol

5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1544-85-0
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.119
MDL Number MFCD00190144
InChI Key CVYQRDKVWVBOFP-UHFFFAOYSA-N
Synonym 2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine
PubChem CID 2736893
IUPAC Name 2,2-difluoro-1,3-benzodioxol-5-amine
SMILES C1=CC2=C(C=C1N)OC(O2)(F)F

Specifications

Boiling Point 95°C to 97°C (12 mmHg)
Odor Recognizable
Refractive Index 1.499
Quantity 5 g
UN Number UN2810
Beilstein 1343593
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 173.12
Percent Purity ≥97%
Chemical Name or Material 5-Amino-2,2-difluoro-1,3-benzodioxole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.