missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2,4-dichloropyrimidine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 5-Amino-2,4-dichloropyrimidine |
| CAS | 5177-27-5 |
| Molecular Formula | C4H3Cl2N3 |
| Quantity | 1 g |
| Synonym | 2,4-Dichloro-5-pyrimidinamine, NSC 86381 |
| InChI Formula | InChI=1S/C4H3Cl2N3/c5-3-2(7)1-8-4(6)9-3/h1H,7H2 |
| SMILES | Nc1cnc(Cl)nc1Cl |
| IUPAC Name | 2,4-dichloropyrimidin-5-amine |
| Molecular Weight (g/mol) | 163.99 |
| Formula Weight | 162.9704 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction